Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LANCL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554063
Description
Immunogen sequence: ALLYLNTEIG PGTVCESAIK EVVNAIIESG KTLSREERKT ERCPLLYQWH RKQYVGAAHG MAGIYYMLMQ PAAKVDQETL TEMVKPSIDY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Necessary for abscisic acid (ABA) binding on the cell membrane and activation of the ABA signaling pathway in granulocytes.
Specifications
LANCL2 | |
Polyclonal | |
Unconjugated | |
LANCL2 | |
1700003F10Rik; G protein-coupled receptor 69B; GPR69B; LanC (bacterial lantibiotic synthetase component C)-like 2; LanC lantibiotic synthetase component C-like 2; LanC lantibiotic synthetase component C-like 2 (bacterial); LanC like 2; LANCL2; lanC-like protein 2; Tasp; Testis-specific adriamycin sensitivity protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55915 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NS86 | |
LANCL2 | |
Recombinant protein corresponding to Human LANCL2. Recombinant protein control fragment (Product #RP-92717). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction