Learn More
Sklar InstrumentsSupplier Diversity Partner Lange Bandage Scissors, Sklar, 7 in., Blunt/Blunt, Straight, Stainless Steel. Sklar's Lange Bandage Scissors are durable, heavy pattern scissors designed for cutting thick dressing material. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 111258
Lange Bandage Scissors, Sklar, 7 in., Blunt/Blunt, Straight, Stainless Steel. Sklar's Lange Bandage Scissors are durable, heavy pattern scissors designed for cutting thick dressing material.
Catalog No. 50-143-7307