Learn More
Sklar InstrumentsSupplier Diversity Partner TC Langenbeck Delicate Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated, Delicate, Pliers, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218081
TC Langenbeck Delicate Needle Holder, Sklar, 7 in., Blunt, Straight, Cross-Serrated, Delicate, Pliers, Tungsten Carbide / Stainless Steel. Sklar's TC Langenbeck Needle Holder is used for delicate and refined suturing. It features plier handles, but has no spring action and no locking mechanism. This instrument is often chosen by users who prefer a quick release of the needle when suturing. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with delicate, cross-serrated jaws, and an overall length of 7 inches.
Catalog No. 50-143-7765