Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Langerin/CD207 Rabbit anti-Human, Clone: LGRN/3136R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Langerin/CD207
Clone LGRN/3136R
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122570
View Documents
Novus Biologicals
NBP30754820UG
20 μg
Each for $159.82
Add to cart
 
NB122569
View Documents
Novus Biologicals
NBP30754810UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human samples. It is validated for ELISA
Specifications

Specifications

Langerin/CD207
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
50489
IgG
Primary
Store at 4°C.
LGRN/3136R
ELISA 2-4 ug/ml
Unconjugated
Rabbit
Human
CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Recombinant fragment (around aa74-213) of human Langerin/CD207 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit