Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanolin, Anhydrous (USP), ACROS Organics™

$281.44

Chemical Identifiers

CAS 8006-54-0
Synonym Wool fat
Chemical Name or Material Lanolin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
AC413000025 SDS Acros Organics
413000025
Glass bottle 2.5kg Each for $281.44
Check Availability
Add to cart
AC413005000 SDS Acros Organics
413005000
Glass bottle 500g N/A N/A N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications

Chemical Identifiers

8006-54-0
Wool fat
Lanolin

Specifications

2.0 ml 0.10 N NaOH max.
Passes Test
0.035% max.
foreign substances: Passes Test
Authentic
Wool fat
0.1% max.
Water-soluble acids and alkalies: Passes Test
38° to 44°C
Lanolin
8006-54-0
Passes Test
Adhering Crystals or Powder
18 to 36g I2/100 g
15, 5409
Passes Test
0.25% max.
Water-soluble oxidizable substances: Passes Test
White to yellow

Safety and Handling

EINECSNumber : 232-348-6

RTECSNumber : OE3201000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit