Learn More
Lanolin, USP, Spectrum™
Click to view available options
Quantity:
430 g
6 x 430 g
18 kg
180 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
8006-54-0
8006-54-0
LA109, 8006-54-0
Spectrum™ Lanolin, USP is used as a moisturizer to treat or prevent dry, rough, scaly, itchy skin and minor skin irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Ignition Residue 0.001
Melting Range 38°-44°C
Quantity 18 kg
CAS Max % 1