Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lansoprazole, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
103577-45-3
C16H14F3N3O2S
369.36
MJIHNNLFOKEZEW-UHFFFAOYNA-N
2-{[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methanesulfinyl}-1H-1,3-benzodiazole
CC1=C(CS(=O)C2=NC3=CC=CC=C3N2)N=CC=C1OCC(F)(F)F
103577-45-3
C16H14F3N3O2S
369.36
MJIHNNLFOKEZEW-UHFFFAOYNA-N
2-{[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methanesulfinyl}-1H-1,3-benzodiazole
CC1=C(CS(=O)C2=NC3=CC=CC=C3N2)N=CC=C1OCC(F)(F)F

L1513, 103577-45-3, C16H14F3N3O2S

Spectrum™ Lansoprazole, USP is used to treat certain stomach and esophagus problems such as acid reflux, ulcers. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 1 g
CAS Max % 1
Percent Purity 98 to 102%