Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lanthanum aluminum oxide, 99.9% (metals basis)

Click to view available options
Quantity:
5g
25g
12003-65-5
MFCD00144550
Lanthanum aluminate
12003-65-5
MFCD00144550
Lanthanum aluminate
Solubility Information Soluble in phosphorous acid (at >150°C). Insoluble in mineral acid ( at 25°C) and water.
Formula Weight 213.88
Physical Form Powder
Percent Purity 99.9%
Odor Odorless
Quantity 5g
Chemical Name or Material Lanthanum aluminum oxide
Assay (metals basis)

EINECSNumber : 234-433-3

TSCA : No

Recommended Storage : Ambient temperatures