Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished Two Sides, 100 Orientation, Alfa Aesar™

100 Orientation

Manufacturer: Alfa Aesar 41434KU

Quantity 5 ea.

 View more versions of this product

Catalog No. AA41434KU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12003-65-5
    Physical Form Plate
    Molecular Formula LaAlO3
    MDL Number MFCD00144550
    Chemical Name or Material Lanthanum Aluminum Oxide Substrate, Polished Two Sides

    Specifications

    Physical Form Plate
    Quantity 5 ea.
    Color Yellow-brown
    MDL Number MFCD00144550
    Odor Odorless
    Molecular Formula LaAlO3
    CAS 12003-65-5
    Chemical Name or Material Lanthanum Aluminum Oxide Substrate, Polished Two Sides

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit