Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Chloride, 7-Hydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Wide Mouth Amber Glass Bottle
10099-58-8
LaCl3·7H2O
10099-58-8
LaCl3·7H2O

High quality chemicals for laboratory and specialized industrial use

    For Flame Enhancement in Atomic Absorption Spectroscopy
Formula Weight 371.38
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 100g
Chemical Name or Material Lanthanum Chloride, 7-Hydrate