Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum chloride heptahydrate, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
10025-84-0
Cl3La·7H2O
371.38
MFCD00149756
10025-84-0
Cl3La·7H2O
371.38
MFCD00149756

Lanthanum chloride heptahydrate, 99.99%, Cl3La.7H2O, CAS Number-10025-84-0, 500g, White, 371.38, Trace Metal Basis, GHS Signal Word: Danger

Lanthanum chloride heptahydrate, 99.99%, Cl3La.7H2O, CAS Number-10025-84-0, 500g, White, 371.38, Trace Metal Basis, GHS Signal Word: Danger

for ultra trace analysis
Assay Percent Range 99.99%
Linear Formula LaCl3·7H2O
Merck Index 15, 5414
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acids and alcohols
Formula Weight 371.38
Physical Form Crystals or Crystalline Powder and/or Chunks
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in acids and alcohols
Total Impurity 0.01% max.
Color White
Melting Point 91.0°C
Quantity 100g
Chemical Name or Material Lanthanum chloride heptahydrate
Show More Show Less

GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger