Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum foil, 1.0mm (0.04in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7048346
Click to view available options
Quantity:
25 x 25 mm
7439-91-0
La
138.91
MFCD00011066
FZLIPJUXYLNCLC-UHFFFAOYSA-N
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
23926
CHEBI:33336
lanthanum
[La]
7439-91-0
La
138.91
MFCD00011066
FZLIPJUXYLNCLC-UHFFFAOYSA-N
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
23926
CHEBI:33336
lanthanum
[La]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Silver to Gray
Quantity 25 x 25 mm
UN Number UN3208
Percent Purity 99.9%
Packaging Mylar under Argon
Weight ≈3.9g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Lanthanum foil, 1.0mm (0.04 in.) thick
Hazard Category H228
Hazard Statement GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
EINECSNumber 231-099-0
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit