Alfa Aesar™ Lanthanum(III) 2-ethylhexanoate, 10% w/v in hexane

Manufacturer: Alfa Aesar™ 41268AC

Quantity 25mL

 View more versions of this product

Catalog No. AA41268AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67816-09-5
    Formula Weight 568.18
    MDL Number MFCD00798555
    UN Number UN1208
    Synonym 2-Ethylhexanoic acid lanthanum(III) salt
    Chemical Name or Material Lanthanum(III) 2-ethylhexanoate

    Specifications

    Formula Weight 568.18
    Health Hazard 2 GHS H Statement
    H225-H304-H361-H373-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P280h-P273-P243-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    Avoid release to the environment.
    Take precautionary measures against static discharge.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.
    UN Number UN1208
    Synonym 2-Ethylhexanoic acid lanthanum(III) salt
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00798555
    CAS 67816-09-5
    Health Hazard 1 Danger
    Quantity 25mL
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
    Concentration 10% w/v in hexane
    Chemical Name or Material Lanthanum(III) 2-ethylhexanoate

    GHS H Statement
    H225-H304-H361-H373-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P280h-P273-P243-P301+P330+P331-P315
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    Avoid release to the environment.
    Take precautionary measures against static discharge.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Get immediate medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit