Learn More
Lanthanum(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Description
Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1,000°C |
Melting Point | 860°C |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
UN Number | UN3077 |
Merck Index | 14,5363 |
Solubility Information | Soluble in water. |
Formula Weight | 245.27 |
Percent Purity | 99.9% |
Show More |
Safety and Handling
Hazard Category | H290-H317-H318 |
Hazard Statement | GHS H Statement H318-H290-H317 Causes serious eye damage. May be corrosive to metals. May cause an allergic skin reaction. |
Precautionary Statement | P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 233-237-5 |
RTECSNumber | OE4375000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.