Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) chloride heptahydrate, 99.99% (REO), Alfa Aesar™, REacton™

Manufacturer:  Alfa Aesar 1126830

Quantity 250g

 View more versions of this product

Catalog No. AA1126830


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10025-84-0
99.99% (REO)
371.37; 245.27 (anhydrous)
Lanthanum(III) chloride heptahydrate
Wet Crystalline Aggregates
LaCl3·7H2O
MFCD00149756

Specifications

10025-84-0
99.99% (REO)
White wet crystalline aggregates
MFCD00149756
White
Lanthanum(III) chloride heptahydrate
Wet Crystalline Aggregates
LaCl3·7H2O
371.37; 245.27 (anhydrous)
Odorless
250g

Safety and Handling

SDS & Safety Documents

H315-H319-H335-H303: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

EINECSNumber : 233-237-5

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit