Lanthanum(III) chloride heptahydrate, ACS

Manufacturer: Alfa Aesar 1291618

Quantity 50g

 View more versions of this product

Catalog No. AA1291618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-84-0
    Molecular Formula LaCl3·7H2O
    Formula Weight 371.37 (245.27 Anhydrous)
    MDL Number MFCD00149756
    Merck Index 14,5363
    Chemical Name or Material Lanthanum(III) chloride heptahydrate, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 371.37 (245.27 Anhydrous)
    Quantity 50g
    TSCA TSCA
    Merck Index 14,5363
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water fo
    RTECS Number OE4470000
    Physical Form Crystalline
    Molecular Formula LaCl3·7H2O
    MDL Number MFCD00149756
    CAS 10025-84-0
    Chemical Name or Material Lanthanum(III) chloride heptahydrate, For analysis ACS
    Grade ACS Grade

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water fo

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit