Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) chloride hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7058723
Click to view available options
Quantity:
100 g
500 g
20211-76-1
Cl3La
245.26
MFCD00149753
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
[Cl-].[Cl-].[Cl-].[La+3]
20211-76-1
Cl3La
245.26
MFCD00149753
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
[Cl-].[Cl-].[Cl-].[La+3]

Lanthanum(III) chloride is useful as precursor for synthesis of aqueous sol-gel lanthanum phosphate nano rods,as a filter aid,an effective flocculent and precipitation phosphate. It is also used as a catalyst for the high pressure oxidative chlorination of methane to chloromethane and organic synthesis it acts as Lewis acid catalyst. It is used as a scintillator material and in Gamma Detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 91°C (decomposition)
pH 5
Physical Form Crystalline
Quantity 100 g
Merck Index 14,5363
Solubility Information Soluble in water and ethanol.
Formula Weight 245.27 (Anhydrous)
Percent Purity 99.9%
Odor Odorless
Assay (REO)
Chemical Name or Material Lanthanum (III) chloride hydrate
Show More Show Less
Hazard Category H290-H317-H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 233-237-5
RTECSNumber OE4470000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit