Alfa Aesar™ Lanthanum(III) chloride, ultra dry, 99.9% (REO)

Manufacturer: Alfa Aesar™ 3570218

Quantity 50g

 View more versions of this product

Catalog No. AA3570218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10099-58-8
    Formula Weight 245.27
    MDL Number MFCD00011068
    UN Number UN3077
    Merck Index 14,5363
    Chemical Name or Material Lanthanum(III) chloride, Ultra dry

    Specifications

    Formula Weight 245.27
    Boiling Point 1,000°C
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P310-P406-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in corrosive resistant container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Hygroscopic
    CAS 10099-58-8
    Melting Point 860°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H290-H317
    Causes serious eye damage.
    May be corrosive to metals.
    May cause an allergic skin reaction.
    RTECS Number OE4375000
    Mesh Size −10 Mesh
    Physical Form Beads
    MDL Number MFCD00011068
    UN Number UN3077
    Merck Index 14,5363
    Density 3.842g/mL
    TSCA TSCA
    Odor Odorless
    Quantity 50g
    Storage Note 1
    • Ampouled under argon
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Color White
    Chemical Name or Material Lanthanum(III) chloride, Ultra dry
    Assay 99.9%, (REO)

    GHS H Statement
    H318-H290-H317
    Causes serious eye damage.
    May be corrosive to metals.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P305+P351+P338-P310-P406-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in corrosive resistant container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit