Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Lanthanum(III) chloride, ultra dry, 99.99% (REO)
Click to view available options
Quantity:
5g
25g
100g
Description
- Ampouled under argon
Specifications
UN Number | UN3077 |
Merck Index | 14,5363 |
Solubility Information | Soluble in water. |
Formula Weight | 245.27 |
Physical Form | Granular,Ampouled under argon |
Percent Purity | 99.99% |
Density | 3.842 g/mL |
Odor | Odorless |
Packaging | Ampule |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
GHS H Statement
H318-H290-H317
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c
H290-H317-H318
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 233-237-5
RTECSNumber : OE4375000
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen