Alfa Aesar™ Lanthanum(III) isopropoxide, La 40%

Manufacturer: Alfa Aesar™ 1461602

Quantity 0.5g

 View more versions of this product

Catalog No. AA1461602

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19446-52-7
    Linear Formula La[OCH(CH3)2]3
    Formula Weight 316.18
    MDL Number MFCD00015305
    UN Number UN2925
    Chemical Name or Material Lanthanum(III) isopropoxide

    Specifications

    Formula Weight 316.18
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    MDL Number MFCD00015305
    UN Number UN2925
    Health Hazard 2 GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    Melting Point 120° to 128°C
    Physical Form Powder
    Linear Formula La[OCH(CH3)2]3
    Health Hazard 1 Danger
    Color White
    Boiling Point 170°C (0.04mmHg) (subl.)
    Quantity 0.5g
    Odor Alcohol-like
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    CAS 19446-52-7
    Concentration La 40%
    Chemical Name or Material Lanthanum(III) isopropoxide

    GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit