Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
10277-43-7
LaN3O9·6H2O
433.01
MFCD00149751
GJKFIJKSBFYMQK-UHFFFAOYSA-N
lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate
16211478
lanthanum(3+);trinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
10277-43-7
LaN3O9·6H2O
433.01
MFCD00149751
GJKFIJKSBFYMQK-UHFFFAOYSA-N
lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate
16211478
lanthanum(3+);trinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[La+3]
For ultra trace analysis
Assay Percent Range 99.995%
Linear Formula La(NO3)3·6H2O
Fieser 11,283
Merck Index 15, 5414
Solubility Information Solubility in water: 1580g/L (25°C). Other solubilities: soluble in acids,soluble in alcohol
Formula Weight 433.01
Physical Form Powder and Granules
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: 1580g/L (25°C). Other solubilities: soluble in acids, soluble in alcohol
Total Trace Metal Impurities 0.005% max
Boiling Point 126.0°C
Color Gray
Melting Point 40.0°C
Quantity 5g
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger