Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) oxide, 99.9% (REO), Alfa Aesar™, REacton™

Manufacturer:  Alfa Aesar 1127222

Quantity 100g

 View more versions of this product

Catalog No. AA1127222


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1312-81-8
99.9% (REO)
325.82
Lanthanum sesquioxide
Powder/Lump
La2O3
MFCD00011071
Lanthanum(III) oxide

Specifications

1312-81-8
99.9% (REO)
White powder/lump
MFCD00011071
6.51g/cm3 at 20°C
White
Lanthanum(III) oxide
Powder/Lump
La2O3
325.82
Lanthanum sesquioxide
Odorless
100g

Safety and Handling

SDS & Safety Documents

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-200-5

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit