Learn More
Lanthanum(III) perchlorate hexahydrate, Thermo Scientific Chemicals
Description
Lanthanum(III) perchlorate hexahydrate participates in the preparation of the Schiff base complexes of lanthanide (III) ions. When hydrated lanthanum perchlorate in ethanolic solution is treated with an ethanolic solution of terpyridine, [La(terpy)3](CIO4)3 is deposited as very small crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN1481 |
Solubility Information | Easily soluble in water. |
Formula Weight | 545.35 (437.26 Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Chemical Name or Material | Lanthanum(III) perchlorate hexahydrate |
Safety and Handling
Hazard Category | H272-H315-H319 |
Hazard Statement | GHS H Statement H272-H315-H319 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S. |
EINECSNumber | 237-835-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.