Alfa Aesar™ Lanthanum Nickel eutectic pieces, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1339918

Quantity 50g

 View more versions of this product

Catalog No. AA1339918

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00198177
    UN Number UN3178
    Chemical Name or Material Lanthanum Nickel eutectic pieces
    Grade Reacton™

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Pieces
    MDL Number MFCD00198177
    UN Number UN3178
    Health Hazard 1 Danger
    Sensitivity Air & Moisture Sensitive
    Melting Point 480°C
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    Quantity 50g
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H350-H228-H317
    May cause cancer.
    Flammable solid.
    May cause an allergic skin reaction.
    Avg. Mol. Wt. or Mol. Wt. Range La:Ni; 88:12 wt%
    Chemical Name or Material Lanthanum Nickel eutectic pieces
    Assay 99.9%, (REO)
    Grade Reacton™

    GHS H Statement
    H350-H228-H317
    May cause cancer.
    Flammable solid.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit