Alfa Aesar™ Lanthanum nickel powder, -100 mesh, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 8913314

Quantity 25g

 View more versions of this product

Catalog No. AA8913314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12196-72-4
    Molecular Formula LaNi5
    MDL Number MFCD00198177
    UN Number UN3178
    Chemical Name or Material Lanthanum nickel powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P231+P232-P261-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 12196-72-4
    Molecular Formula LaNi5
    MDL Number MFCD00198177
    UN Number UN3178
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H260-H350-H317
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.
    May cause cancer.
    May cause an allergic skin reaction.
    TSCA TSCA
    Physical Form Intermetallic
    Mesh Size −100 Mesh
    Sensitivity Air & Moisture Sensitive
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    Quantity 25g
    Odor Odorless
    Chemical Name or Material Lanthanum nickel powder
    Assay 99.5%, (metals basis)

    GHS H Statement
    H228-H260-H350-H317
    Flammable solid.
    In contact with water releases flammable gases which may ignite spontaneously.
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P231+P232-P261-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit