Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Oxide, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
250g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

1312-81-8
La2O3

High quality chemicals for laboratory and specialized industrial use

CAS 1312-81-8
Molecular Formula La2O3
Formula Weight 325.81
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 6.51kg
Quantity 100g
Chemical Name or Material Lanthanum Oxide
Grade BAKER ANALYZED™ Reagent