Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Oxide, Reagents

Catalog No. p-200021199
Click to view available options
Quantity:
100 g
500 g
1 kg
10 kg

Lanthanum Oxide, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
Molecular Weight (g/mol) 325.81
InChI Key MRELNEQAGSRDBK-UHFFFAOYSA-N
Synonym Lanthanum sesqioxide
IUPAC Name dilanthanum(3+) trioxidandiide
SMILES [O--].[O--].[O--].[La+3].[La+3]

Specifications

Boiling Point 4200°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 6.51
Chemical Name or Material Lanthanum Oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.