Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13870AP

Quantity 500mL

 View more versions of this product

Catalog No. AA13870AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7697-37-2,1312-81-8,7732-18-5
La2O3 in 5% HNO3
Lanthanum
Liquid
MFCD00011066

Specifications

7697-37-2,1312-81-8,7732-18-5
Liquid
MFCD00011066
23 hPa (17mm Hg) at 20°C
500mL
Clear liquid
La2O3 in 5% HNO3
Nitric acid: 5.0%; Lanthanum(III) oxide: 0.12%; Water: 94.88%
Clear colorless
Lanthanum

Safety and Handling

SDS & Safety Documents

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit