Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Rod, 6.35mm (0.25 in.) dia., 99.9% (REO), Alfa Aesar™

Form: Approx.0.20g/mm, Cast rod, diameter is approximate

Manufacturer:  Alfa Aesar 40292HF

Quantity 10mm

 View more versions of this product

Catalog No. AA40292HF


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7439-91-0
99.9% (REO)
Lanthanum, Rare Earth Oxides Content
Rod, 6.35mm (0.25 in.) dia.
MFCD00011066

Specifications

10mm
Silvery-white rod
99.9% (REO)
Silver-white
7439-91-0
Rod, 6.35mm (0.25 in.) dia.
MFCD00011066
Lanthanum, Rare Earth Oxides Content

Safety and Handling

SDS & Safety Documents

Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.

H228-H261: Flammable solid. In contact with water releases flammable gas.

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 231-099-0

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit