Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lanthanum Solution, 5% w/v as La, Reagents
Click to view available options
Quantity:
500 mL
4 L
20 L
Specifications
Specifications
Boiling Point | 100°C |
Chemical Name or Material | Lanthanum Solution, 5% w/v as La |
Color | Colorless |
CAS | 7732-18-5, 7647-01-0 |
Physical Form | Liquid |
Quantity | 4 L |
Synonym | Lanthanum sesqioxide |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.1035 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction