Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lanthanum sputtering target, 76.2mm dia x 1.59mm thick

Manufacturer:  Alfa Aesar™ 46064KS

Quantity 1 each

 View more versions of this product

Catalog No. AA46064KS


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7439-91-0
MFCD00011066
UN3208
Lanthanum sputtering target, 76.2mm dia. x 1.59mm thick

Specifications

1 each
MFCD00011066
Lanthanum sputtering target, 76.2mm dia. x 1.59mm thick
7439-91-0
UN3208

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit