Alfa Aesar™ Lanthanum titanium oxide

Manufacturer: Alfa Aesar™ 1821322

Quantity 100g

 View more versions of this product

Catalog No. AA1821322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12031-47-9
    Formula Weight 485.57
    MDL Number MFCD00016167
    Synonym Lanthanum titanate
    Chemical Name or Material Lanthanum titanium oxide

    Specifications

    Formula Weight 485.57
    Odor Odorless
    CAS 12031-47-9
    Synonym Lanthanum titanate
    Color White
    Quantity 100g
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00016167
    Chemical Name or Material Lanthanum titanium oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit