Learn More
LanthaScreen™ Eu-anti-DYKDDDDK Antibody
LanthaScreen™ Eu-anti-DYKDDDDK Antibody
$912.00
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
PV6026 Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
PV6026
N/A 25μg
Each for $912.00
Add to cart
Description
Description
LanthaScreen Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity.
Safety and Handling
Safety and Handling
SDS
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.