Learn More
LanthaScreen™ Eu-PY20 Antibody Kit
LanthaScreen™ Eu-PY20 Antibody Kit
$1052.00
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
PV5692 Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
PV5692
N/A 25μg
Each for $1,052.00
Add to cart
Description
Description
A general reagent to monitor tyrosine kinase activity, the PY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate. For use in combination with AlexaFluor 647-poly-GT (PV5693) to evaluate inhibitors in medium to high throughput screening applications.
Safety and Handling
Safety and Handling
SDS
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.