Learn More
LanthaScreen™ Tb-anti-Biotin Antibody
LanthaScreen™ Tb-anti-Biotin Antibody
$906.00
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
PV4745 Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
PV4745
N/A 25μg
Each for $906.00
Add to cart
Description
Description
A general reagent for use in assay development, the anti-biotin antibody binds to biotinylated peptide or physiological protein substrates.
Safety and Handling
Safety and Handling
SDS
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.