Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LanthaScreen™ Tb-anti-GST Antibody

Encompass

LanthaScreen™ Tb-anti-GST (Goat) Antibody

$994.00

Specifications

Concentration 0.55mg/mL
Molecular Weight of Antigen 150kDa
Packaging 1 Each
Primary or Secondary Primary
Storage Requirements –20°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3550
Encompass
View Documents Promotion Details
LIFE TECHNOLOGIES
PV3550
25μg
Each for $994.00
Add to cart
 
Description

Description

LanthaScreen™ terbium-labeled anti-GST Goat antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity.
Specifications

Specifications

0.55mg/mL
1 Each
–20°C
Tb (Terbium)
RUO
150kDa
Primary
Drug Discovery and Development, Immunoprecipitation, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Protein Purification and Isolation, Target and Lead Identification and Validation, TR-FET Assay Development, Western Blot
Liquid
Primary
Safety and Handling

Safety and Handling

SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit