Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LanthaScreen™ Tb-pATF2 (pThr71) Antibody Non-distribution product as customer accommodation.

LanthaScreen™ Tb-pATF2 (pThr71) Antibody

$18190.00

Specifications

Packaging 1 Each
Primary or Secondary Primary
Storage Requirements –20°C
Conjugate Tb (Terbium)
Format Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV4448 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV4448
1mg Each for $18,190.00
Add to cart
 
Description

Description

A member of the leucine zipper family of DNA binding proteins, ATF2 (activating transcription factor 2) is phosphorylated at Thr71 by members of the JNK and p38 families of kinases.

For use with the GFP-ATF2 physiological protein substrate (PV4445) to evaluate inhibitors in medium to high throughput screening applications.
Specifications

Specifications

1 Each
–20°C
Liquid
Primary
Primary
Tb (Terbium)
RUO, For research use only

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit