Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LanthaScreen™ Tb-PY20 Antibody Non-distribution product as customer accommodation.

LanthaScreen™ Tb-PY20 Antibody

$9120.00

Specifications

Antigen Tyrosine kinases
Concentration 0.54mg/mL
Molecular Weight of Antigen 150kDa
Packaging 1 Each
Primary or Secondary Primary
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3529 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV3529
1mg Each for $9,120.00
Add to cart
 
Description

Description

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.
For use in combination with Fluorescein-poly-GT (PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.
Specifications

Specifications

Tyrosine kinases
150kDa
Primary
Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology, Pharma and Biopharma, Target and Lead Identification and Validation
RUO, For research use only
0.54mg/mL
1 Each
–20°C
Tb (Terbium), lanthanide chelates
Primary

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit