Learn More
Invitrogen™ LanthaScreen™ Tb-Streptavidin, 1 mg/mL Kit
LanthaScreen™ Tb-Streptavidin, 1 mg/mL Kit
$1782.00
Specifications
Concentration | 1 mg/mL |
---|---|
Conjugate | Tb (Terbium) |
Content And Storage | LanthaScreen™ Tb-Streptavidin is supplied at 1 mg/ml and contains the streptavidin in HEPES buffered saline, 400 µl of the fluorescein-labeled, biotinylated peptide for use as a positive control, and 100 ml of TR-FRET Dilution Buffer. Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C). |
Shipping Condition | Dry Ice |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PV3965
![]() |
Invitrogen™
PV3965 |
50 μg |
Each of 1 for $1,782.00
|
|
|||||
Description
As part of the LanthaScreen™ TR-FRET Toolbox of assay reagents, LanthaScreen™ Tb-labeled streptavidin is a generic reagent that can be used to easily label biotinylated biomolecules with terbium for use in TR-FRET assays. Although the terbium chelate donor can be directly attached to a biomolecule such as a protein or peptide, the Tb-labeled streptavidin can be indirectly attached to a target molecule through a biotin-mediated association with Tb-labeled streptavidin.Specifications
1 mg/mL | |
LanthaScreen™ Tb-Streptavidin is supplied at 1 mg/ml and contains the streptavidin in HEPES buffered saline, 400 µl of the fluorescein-labeled, biotinylated peptide for use as a positive control, and 100 ml of TR-FRET Dilution Buffer. Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C). | |
Liquid | |
Streptavidin Conjugate (Fluorescent) |
Tb (Terbium) | |
Dry Ice | |
LanthaScreen™ | |
Tb (Terbium) |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.