Learn More
Lanx Chemical Protective Boot Liner
Durable with outstanding vapor, aerosol, and fire protection
Manufacturer:  Lanx CPUSOCKSM
 View more versions of this product
Catalog No. 19-130-0236B
Add to cart
Description
Description
  • Unique stretch charteristics offer form-fitting comfort and minimizes sizing tariff
  • Air permeable to reduce heat stress via evaporative cooling
  • Mid-calf design integreates with most footwear
Order Info
Can be laundered multiple times on gentle cyle in non-phosphate detergents and tumble dried on low.
Compliance
Lanx Type I fabric meets U.S. military specification MIL-U-44435.
Specifications
Specifications
Reduce heat stress
Composite Fabric
MIL-U-44435
Medium