Learn More
Lanx Chemical Protective Integrated Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Cotton liner
- Butyl rubber with leather palm
- Ideal for chemical and biological applications
- Vacuum packed
Specifications
Color | Black |
Size | Small |