Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAP (TGF-beta 1) Antibody (27232), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2463L
This item is not returnable.
View return policy
Description
LAP (TGF-beta 1) Monoclonal antibody specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Flow CytometrySpecifications
| LAP (TGF-beta 1) | |
| Monoclonal | |
| DyLight 550 | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-beta 1), Leu30-Arg278 (Cys33Ser), Accession # P01137 | |
| 0.5 mL | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 7040 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 27232 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction