β-Lapachone, Tocris

DNA topoisomerase I inhibitor

Manufacturer: R&D Systems 1293/10

Quantity 10mg

 View more versions of this product

Catalog No. 12-931-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C15H14O3
    Formula Weight 242.27
    Synonym 3,4-Dihydro-2,2-dimethyl-2H-naphtho[1,2-b]pyran-5,6-dione
    Chemical Name or Material β-Lapachone

    Specifications

    Formula Weight 242.27
    Quantity 10mg
    Synonym 3,4-Dihydro-2,2-dimethyl-2H-naphtho[1,2-b]pyran-5,6-dione
    Assay Percent Range >98%
    Molecular Formula C15H14O3
    CAS 4707-32-8
    Recommended Storage Store at +4°C
    Chemical Name or Material β-Lapachone

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit