Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LAPTM4B Monoclonal Antibody (CL5287)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531416
Description
Immunogen sequence: DTTVLLPPYD DATVNGAAKE.
LAPTM4B is a multi-pass membrane protein. It belongs to the LAPTM4/LAPTM5 transporter family. LAPTM4b has active role in disease progression of malignant cells and is involved in cell proliferation and multidrug resistance. The genetic polymorphism of LAPTM4B is a potential risk factor for the development of colon cancer and gastric cancer. The research results also indicated that LAPTM4B may be a clinically useful prognostic indicator for ovarian carcinoma and may play a role in human hepatocellular carcinoma.
Specifications
LAPTM4B | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86VI4 | |
LAPTM4B | |
Synthetic peptide corresponding to Human LAPTM4B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin) | |
CL5287 | |
Unconjugated | |
LAPTM4B | |
C330023P13Rik; hypothetical protein 5; Laptm4b; LAPTM4beta; LC27; LOW QUALITY PROTEIN: lysosomal-associated transmembrane protein 4B; lysosomal associated protein transmembrane 4 beta; lysosomal protein transmembrane 4 beta; lysosomal-associated protein transmembrane 4B; lysosomal-associated transmembrane protein 4; lysosomal-associated transmembrane protein 4B; Lysosomal-associated transmembrane protein 4B-like protein; lysosome-associated transmembrane protein 4-beta; PSEC0001 | |
Mouse | |
Protein A | |
RUO | |
55353 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction