Learn More
Invitrogen™ LAPTM4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114204
Description
Immunogen sequence: SCYRYINGRN SSDVLVYVTS NDTTVLLPPY DDATAVPSTA KEPPPPYVSA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 86%.
LAPTM4B is a multi-pass membrane protein. It belongs to the LAPTM4/LAPTM5 transporter family. LAPTM4b has active role in disease progression of malignant cells and is involved in cell proliferation and multidrug resistance. The genetic polymorphism of LAPTM4B is a potential risk factor for the development of colon cancer and gastric cancer. The research results also indicated that LAPTM4B may be a clinically useful prognostic indicator for ovarian carcinoma and may play a role in human hepatocellular carcinoma.
Specifications
LAPTM4B | |
Polyclonal | |
Unconjugated | |
LAPTM4B | |
C330023P13Rik; hypothetical protein 5; Laptm4b; LAPTM4beta; LC27; LOW QUALITY PROTEIN: lysosomal-associated transmembrane protein 4B; lysosomal associated protein transmembrane 4 beta; lysosomal protein transmembrane 4 beta; lysosomal-associated protein transmembrane 4B; lysosomal-associated transmembrane protein 4; lysosomal-associated transmembrane protein 4B; Lysosomal-associated transmembrane protein 4B-like protein; lysosome-associated transmembrane protein 4-beta; PSEC0001 | |
Rabbit | |
Affinity chromatography | |
RUO | |
114128 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q91XQ6 | |
LAPTM4B | |
Synthetic peptide directed towards the C-terminal region of Mouse Laptm4b (aa 178-227). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.