Learn More
LAR/PTPRF Mouse anti-Human, Mouse, Rat, Biotin, Clone: S165-38, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242172B
Catalog No. NBP242172B
Add to cart
Description
Description
LAR/PTPRF Monoclonal antibody specifically detects LAR/PTPRF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
LAR/PTPRF
S165-38
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PTPRF
Fusion protein amino acids 1315-1607 (cytoplasmic C-terminus) of human LAR. 97% identical in both rat and mouse. >80% identity with PTPRD and PTPRS. >50% identity with PTPRM and PTPRK.
Protein G purified
Store at 4C in the dark.
Monoclonal
Greater then 80% identity with PTPRD and PTPRS. Greater then 50% identity with PTPRM and PTPRK.Detects approx 85kDa (full length protein is 210 kDa -smaller due to proteolysis into P-subunit containing transmembrane and intracellular domains.
Western Blot, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
LCA-homolog, protein tyrosine phosphatase, receptor type, F, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F
Mouse
IgG2a
0.1 ml
Primary
5792
Human, Mouse, Rat