Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LAR4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569095
Description
This target displays homology in the following species: Cow: 85%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 85%; Pig: 86%; Rabbit: 86%; Rat: 92%.
The LAR4B gene, more accurately referred to as LARP4B (La Ribonucleoprotein Domain Family Member 4B), is associated with RNA-binding proteins that are crucial for various aspects of RNA metabolism, including RNA stabilization, translation, and cellular stress responses. LARP4B belongs to the La-related protein family, which plays significant roles in maintaining RNA stability and enhancing translation efficiency of certain mRNAs. It has been identified as a target in cancer research due to its involvement in tumor biology, particularly regulating tumor suppressive pathways and interacting with microRNA networks. Studies have shown that LARP4B expression is downregulated in prostate cancer tissues, indicating its potential role as a tumor suppressor. Moreover, it is linked with liver cancer and survival rates, reflecting its importance in cellular growth and proliferation. The regulation of LARP4B and its interactions with other cellular components underline its potential therapeutic implications for cancer treatment.
Specifications
LAR4B | |
Polyclonal | |
Unconjugated | |
LARP4B | |
A630096F19; AI256361; D13Wsu64e; KIAA0217; La ribonucleoprotein 4B; La ribonucleoprotein domain family member 4B; La ribonucleoprotein domain family member 5; La ribonucleoprotein domain family, member 4B; La ribonucleoprotein domain family, member 5; la-related protein 4B; La-related protein 5; LARP4B; Larp5; RGD1310713 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
217980 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6A0A2 | |
LARP4B | |
synthetic peptide directed towards the middle region of mouse LAR4B. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction