Learn More
Stoelting Large Animal Adaptor
Accommodate a wide size range of large animals in either the Lab Standard™ or Parallel Rail series of stereotaxic instruments
Manufacturer:  Stoelting 51852
 View more versions of this product
Catalog No. 10-000-226
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Dog Adaptor available only for the Parallel Rail (51800) series stereotaxic instruments. To fit any size dog, it should be used with 51850 and 51851 stereotaxic instruments, which have two sets of ear bar holders at different spacings from the front, to accommodate a wide size range of dogs.
Cat Adaptor is available for the Lab Standard™ or Parallel Rail series of stereotaxic instruments. The adaptor for the Lab Standard™ series (51600) includes spacers and bolts to raise the U frame 2.5 inches above the base plate, the eye socket/palate holder and 18° ear bars. Also available with  non-puncture 45° ear bars (51827).
Specifications
Specifications
Neuroscience
Dog
Adaptor