Learn More
Fisherbrand™ Large Bore Male Luer with 5/16 in. ID - Natural Kynar - QC
SDP
Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Large bore luers offer a higher flow rates than standard luers. 
Catalog No. 01-000-182
Add to cart
Description
Description
  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • RoHS Compliant
  • Gamma Stable
  • Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Specifications
Compliance Certificates Available
Natural Kynar
Medical, Pharmaceutical, Industrial applications
Out of direct sunlight, >20°F <100°F
Class 7 Cleanroom Manufactured
Large Bore Luer
100 Pack
Natural