Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LARGE Mouse anti-Human, Alexa Fluor™ 488, Clone: LARGE-02, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP262211AF488

Catalog No. NB002517


Add to cart

Description

Description

LARGE Monoclonal specifically detects LARGE in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

LARGE
LARGE-02
Western Blot, Flow Cytometry
Purified, Conjugated
LARGE
Recombinant fragment of human LARGE1 (amino acids 35-142)
Protein A purified
Store at 4°C in the dark.
Monoclonal
The antibody MEM-65 recognizes an unique epitope of CD2, a 50 kDa glycoprotein present on the human peripheral blood T-lymphocytes and NK cells; also expressed by all thymocytes.
Western Blot, Flow Cytometry
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Acetylglucosaminyltransferase-like 1A, EC 2.4, glycosyltransferase-like protein LARGE1, KIAA0609acetylglucosaminyltransferase-like protein, LARGE1, like-acetylglucosaminyltransferase, like-glycosyltransferase, MDC1D, MDDGA6, MDDGB6
Mouse
IgG2b
0.1 mL
Primary
9215
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit